missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 0.01 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction
Unit Size:
1 set
3mg
600µg
60µg
Description
R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from cyanobacteria, red algae and cryptomonads. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield. It has a strong absorption peak at 565 nm, and a secondary absorption peak at 492 nm. The emission peak is at 575. The amount of antibody used for labeling should be the same as the pack size of LL-R-PE. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 3 x 0.01 mg Reaction |
| Conjugate | PE |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
| Product Type | R-PE Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction