missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 0.01 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction
Unit Size:
1 set
3mg
600µg
60µg
Description
R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 1 x 100 μg Reaction |
| Conjugate | PE |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
| Product Type | R-PE Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction