Learn More
Gibco™ Essential 8™ Flex Medium Kit
Código de producto. 15374799
Click to view available options
Quantity:
1 Kit
Tamaño de la unidad:
1 set
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15374799
Marca: Gibco™ A2858501
Este artículo no se puede devolver. Vea la política de devoluciones
Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) with a flexible feeding schedule.
Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) without the need for weekend feeding. It is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement).
Essential 8 Flex provides all of the benefits of Essential 8 Medium: minimized variability of components, reliable and robust cultures with a xeno-free, cGMP produced medium, along with the added benefit of flexibility. Maintenance of standard PSCs no longer requires being in the lab seven days a week.
Flexibility
With Essential 8 Flex Medium, pluripotent stem cells (PSCs) need only be fed on the day of splitting and the day after. If the cells will be left without feeding for more than 48 hours, such as over a weekend, then simply double the feed volume on Friday.
Reliable and robust cultures
Essential 8 Flex Medium has been shown to support PSC growth for >30 passages without any signs of karyotypic abnormalities and to maintain the ability of iPSCs to differentiate into all three germ line lineages.
Cost effective
Essential 8 Flex Medium is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement), and when used with vitronectin (VTN-N), provides a cost effective, defined system for feeder-free culture of human pluripotent stem cells.
The Essential 8 Flex Medium formulation is based upon that published for Essential 8 Medium by Chen, et al., and allows for control and flexibility of PSC culture medium, without a negative impact to the health of the stem cells.
Información de órdenes
Approved for shipment at Room Temperature or on Wet Ice
TRUSTED_SUSTAINABILITY
Especificaciones
Cell Line Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Xeno-free
Concentration Essential 8 Flex Basal Medium (1X), Essential 8 Flex Supplement (50X)
Form Liquid
Product Type Stem Cell Media
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Product Line Essential 8
Quantity 1 Kit
Shipping Condition Supplement - dry ice, Basal - ambient
Content And Storage • 500 mL Basal Medium (store at 2–8°C and protect from light)
• 10 mL Supplement (store at -5 to -20°C and protect from light)
Culture Type Adherent Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
Mostrar más Mostrar menos
For Research Use Only. Not for use in diagnostic procedures.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado