missing translation for 'onlineSavingsMsg'
Learn More

Lithocholic acid, 95%

Product Code. 10511354 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10511354

Brand: Thermo Scientific Acros 229090250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lithocholic acid, CAS # 434-13-9, a bacterial metabolite, is an oligo (ethylene glycol) and a secondary bile acid. | CAS: 434-13-9 | C24H40O3 | 376.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Lithocholic acid is a monohydroxy acid, and a secondary bile acid

Applications

  • Lithocholic acid can be used in studies that assess cholestasis
  • It can be used to study hepatic phospholipid regulation and homeostasis of bile acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-13-9
Molecular Formula C24H40O3
Molecular Weight (g/mol) 376.581
MDL Number MFCD00003682
InChI Key SMEROWZSTRWXGI-HVATVPOCSA-N
Synonym lithocholic acid, lithocolic acid, lithocholate, 3alpha-hydroxy-5beta-cholanic acid, 3alpha-hydroxy-5beta-cholan-24-oic acid, 3-hydroxycholan-24-oic acid, 3alpha-hydroxycholanic acid, 3-alpha-hydroxycholanic acid, 5beta-cholanic acid-3alpha-ol, 3alpha-hydroxy-5beta-cholanoic acid
PubChem CID 9903
ChEBI CHEBI:16325
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CCC4C3(CCC(C4)O)C)C

Specifications

Melting Point 182°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 10, IV, 785
Merck Index 15, 5601
Specific Rotation On dry basis: +32° to +36° (20°C, 589nm) (c=1.5, C2H5OH)
Formula Weight 376.57
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Lithocholic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.