Learn More
Notrax Logo Standard™ Affordable Absorbent Printed Logo Mat
Click to view available options
Dimensions (L x W):
90 x 60 cm
120 x 90 cm
150 x 90 cm
300 x 90 cm
180 x 120 cm
Unit Size:
Each
Description
- Logo Standard™ is the ideal promotional matting solution for light duty applications
- Using BrilliantStep™ full color printing with almost infinite color combinations, the even velour surface allows fine printing and true color presentation
- A durable vinyl backing with smooth finish adapts to any surface
- Logo Standard™ can be fully customized with BrilliantStep™ state-of-the-art printing technology allowing 3-dimensional effects, fine lines and color shading, for each mat to have its truly individual image
Specifications
Specifications
| Dimensions (L x W) | 180 x 120 cm |
| Style | Frieze |
| Color | Logo |
| Length (English) | 71 in. |
| Length (Metric) | 180 cm |
| Material | Polyamide 6.6 High Twist Yarns |
| Type | Industrial and Commercial Matting |
| Width (Metric) | 120 cm |