Learn More
Cole-Parmer™ Long Rigid Nylon Scraper Spatula
Product Code. 11785269 Shop All Cole Parmer Products
Click to view available options
Length (Metric):
34.29 cm
Unit Size:
Each
Product Code. 11785269
Brand: Cole-Parmer™ 0626610
Only null left
Add to basket
Add to basket
Ideal forhard-to-clean containers
  • Short, comfortable handle and wide nylon blade make these scraper spatulas ideal for hard-to-clean containers
  • Rigid blade for increased strength
  • Model 06266-15 has a soft blade for greater flexibility
Specifications
Product Type Scraper Spatula
Length (English) 13.5 in.
Width (English) 4.5 in.
Width (Metric) 11.43 cm
Material Nylon
Length (Metric) 34.29 cm