Learn More
Lonza™ BioWhittaker™ 293 Cell Serum-Free Media
Completely balanced to support vaccine and viral vector production of high-density human embryonic kidney (HEK) 293 cell culture environments
Brand: Lonza™ BE12-764Q
This item is not returnable.
View return policy
Description
Very low levels of recombinant proteins facilitate downstream purification and provide regulatory compliance
- Chemically-defined media optimized for specific cell types
† Store as detailed on label.
Specifications
2° to 8°C | |
No Glutamine, No Phenol Red | |
For Adherent Cells |
Pro293A-CDM | |
Bottle | |
1 L |