Learn More
Lonza™ BioWhittaker™ Glasgow Minimum Essential Medium (G-MEM, BHK-21)
Brand:  Lonza™ BE12-739F
Product Code. 11695240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Manufactured using formulations recommended by the Tissue Culture Association and Water for Injection (WFI) quality water
  • Sterile-filtered through two consecutive 0.2μm pharmaceutical-grade filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Sterile
7 to 7.4
G-MEM, BHK-21
Sterile-filtered through two consecutive 0.2 μm filters
500 mL