missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker Insect-XPRESS Insect Cell Culture Media (1X) with L-Glutamine
Designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21). BioWhittaker™ Insect-XPRESS™ Insect Cell Culture Media (1X) with L-Glutamine supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS).
95.64 EUR - 124.44 EUR
Specifications
Content And Storage | 2° to 8°C |
---|---|
Form | Liquid |
Product Type | Insect Cell Medium, Protein-free |
With Additives | Glutamine |
Product Code | Brand | Product Type | Quantity | With Additives | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | With Additives | Price | Quantity & Availability | |||
10564038
|
Lonza™
BE12-730Q |
Insect Cell Medium, Protein-free | 1 L | Glutamine |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11635240
|
Lonza™
BE12-730F |
Insect Cell Medium, Protein-free | 500 mL | Glutamine |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Protein-free
- Cell densities in excess of 8.3×106 cells/mL can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and excess of oxygen
- Formulation can also be used for stationary mono¬layer cultures and shake-flask cultures
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
2° to 8°C | |
Liquid | |
Insect Cell Medium, Protein-free | |
Glutamine |