missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ McCoy's 5A Medium (Modified)
General purpose medium for primary and established cell lines. BioWhittaker™ McCoy's 5A Medium (Modified) is manufactured in accordance with cGMP regulations.
28.38 EUR
Specifications
Content And Storage | 2° to 8°C |
---|---|
Product Type | McCoy's 5A |
Sterility | Sterile |
Sterilization Method | Sterile-filtered through two consecutive 0.2 μm filters |
Quantity | 500 mL |
Product Code | Brand | pH | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Price | Quantity & Availability | |||||
11645210
|
Lonza™
BE12-688F |
6.89 to 7.2 | |||||||
Description
- Formulated for the nutritional requirements of Walker 256 carcinoma
- Unique components include asparagine, 3X normal glucose, amino acids (all L-isomers) and increased folic acid
- Manufactured in accordance with cGMP regulations
- Formulations used for standard liquid media products are those recommended by Tissue Culture Association
Specifications
2° to 8°C | |
Sterile | |
500 mL |
McCoy's 5A | |
Sterile-filtered through two consecutive 0.2 μm filters |