Learn More
Lonza™ BioWhittaker™ Medium 199 with Earle's BSS
Brand:  Lonza™ BE12-117F
 View more versions of this product
Product Code. 11635170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Uses include virology and vaccine production
  • Components include purines (adenine, guanine, xanthine, hypoxanthine), pyrimdines (thymine, uracil) and fat soluble compounds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Sterile
Glutamine, HEPES
500 mL
M199/EBSS
Sterile-filtered through two consecutive 0.2 μm filters
6.9 to 7.3