Learn More
Lonza™ BioWhittaker™ Medium 199 with Hanks' BSS
Brand: Lonza™ BE12-109F
This item is not returnable.
View return policy
Description
- Uses include virology and vaccine production
- Components include purines (adenine, guanine, xanthine, hypoxanthine), pyrimdines (thymine, uracil) and fat soluble compounds
Specifications
2° to 8°C | |
Sterile | |
7.08 to 7.4 |
M199/HBSS | |
Sterile-filtered through two consecutive 0.2 μm filters | |
500 mL |