missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ MEM Eagle with Earle's BSS (10X)

Brand:  Lonza™ BE12-684F

Product Code. 11635210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

MEM Eagle is suitable for a diverse spectrum of mammalian cell types.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Sterile
No Glutamine, No Sodium Bicarbonate
500 mL
MEM Eagle/EBSS
Sterile-filtered through two consecutive 0.2 μm filters
5.2 to 6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.