missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ MEM Eagle Nonessential Amino Acid Solution (100X)
Used to enhance cell growth. BioWhittaker™ MEM Eagle Nonessential Amino Acid Solution (100X) is manufactured in accordance with cGMP regulations.
Brand: Lonza™ BE13-114E
This item is not returnable.
View return policy
Description
- Manufactured under cGMP regulations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
2° to 8°C | |
Amino Acid Supplement | |
2.15 to 3.35 |
Liquid | |
Sterile | |
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction