Learn More
Lonza™ BioWhittaker™ Sodium Pyruvate Solution (100mM)
Added to cell culture as an additional source of energy since it is a key intermediate in the production of the high energy intermediate ATP within the cell BioWhittaker™ Sodium Pyruvate Solution (100mM) is manufactured in accordance with cGMP regulations.
Brand: Lonza™ BE13-115E
This item is not returnable.
View return policy
Description
- Sodium Pyruvate Solution may help cells that are deficient in the nine enzymatic steps that break glucose down to pyruvate.
- May also assist in “jump starting” cells in the citric acid cycle and thus help low density cells grow and produce energy in the form of ATP
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
2° to 8°C | |
Sodium Pyruvate | |
100 mL |
Liquid | |
Sterile |