Learn More
Lonza™ BioWhittaker™ TrypZean™ EDTA Solution
Solution that is of completely non-animal origin, intended for cell dissociation
Brand: Lonza™ BE02-034E
This item is not returnable.
View return policy
Description
- NAO trypsin for adherent cell lines: produced in a plant-based system, without using any animal origin components
- Ready to use
- Active ingredient is TrypZean, a recombinant bovine trypsin expressed in corn
- TrypZean solution is supplemented with EDTA, which helps for cell detachment by catching CA and MG ions
- Convenient for serum-free and serum supplemented cell cultures
- No protocol change for users of porcine trypsin
- Inhibition of TrypZean has to be performed the same way as for the native form, using either serum or classical inhibitors
- pH range for use is 7.1 to 7.9
- Solution contains phenol red, allowing for detection of pH shift
Specifications
100 mL |