Learn More
Lonza™ BioWhittaker™ UltraMEM Reduced-Serum Media
For supporting the growth and maintenance of anchorage-dependent cells
Brand: Lonza™ BE12-743F
This item is not returnable.
View return policy
Description
- Support growth and maintenance of several anchorage-dependent cell types under reduced serum concentrations
- Use with 0 to 4% serum
- Growth performance can be increased by the addition of insulin, transferrin, selenium and ethanolamine
Specifications
UltraMEM Reduced-Serum Media | |
Sterile | |
7.2 to 7.6 | |
500 mL |