Learn More
Lonza™ DMEM:F12; 1:1 Mixture
Used to demonstrate the effect of various hormones and growth factors on target tissues. Lonza™ DMEM:F12; 1:1 Mixture is also well-suited for clonal density cultures.
Brand:  Lonza™ BE04-687F/U1
Product Code. 11625140
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Combines the richness of F12 with the higher component concentrations of DMEM
  • Formulation used for this standard liquid media product is recommended by the Tissue Culture Association
  • Has been extensively used to demonstrate the effect of various hormones and growth factors on target tissues
  • For further manufacturing or laboratory use
  • These products are for research use only
  • Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Liquid
DMEM:F12; 1:1 Mixture
500 mL