Learn More
Lonza™ DMEM:F12; 1:1 Mixture
Used to demonstrate the effect of various hormones and growth factors on target tissues. Lonza™ DMEM:F12; 1:1 Mixture is also well-suited for clonal density cultures.
Brand: Lonza™ BE04-687F/U1
This item is not returnable.
View return policy
Description
- Combines the richness of F12 with the higher component concentrations of DMEM
- Formulation used for this standard liquid media product is recommended by the Tissue Culture Association
- Has been extensively used to demonstrate the effect of various hormones and growth factors on target tissues
- For further manufacturing or laboratory use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Specifications
2° to 8°C | |
Liquid | |
DMEM:F12; 1:1 Mixture | |
500 mL |