missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System Components and Packs: FlashGel Camera

Fast analysis, recovery, and documentation of DNA and RNA

Brand:  Lonza™ 57040

Product Code. 11537865

  • 1840.00 EUR / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.

This item is not returnable. View return policy

Description

Description

  • Provides nearly instant results
  • Load samples, watch bands migrate and get data in as little as two minutes
  • Includes DNA recovery and gel documentation
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Clean, sharp separation and straight, uniform sample lanes

FlashGel Camera

  • Includes: Camera, hood enclosure, USB cable and software installation CD
  • Compact design; hood measures 10L x 11W x 16cm H (3.9 x 4.3 x 6.3 in.)
  • For use with the FlashGel Dock (Cat. No. BMA57025)

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

TRUSTED_SUSTAINABILITY
Specifications

Specifications

FlashGel Camera
FlashGel Camera, Hood Enclosure, USB Cable and Software Installation CD
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.