Learn More
Lonza™ FlashGel™ System Components and Packs: FlashGel Dock
Fast analysis, recovery, and documentation of DNA and RNA
Brand:  Lonza™ 57025
Product Code. 11547855
  • 1390.00 EUR / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Alert:
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Description
Description
  • Provides nearly instant results
  • Load samples, watch bands migrate and get data in as little as two minutes
  • Includes DNA recovery and gel documentation
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Clean, sharp separation and straight, uniform sample lanes
FlashGel Dock
  • For use with all FlashGel cassettetypes
  • Provides electrophoresis and visualization of both DNA and RNA cassettes
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FlashGel Dock
FlashGel Dock