Learn More
Lonza™ FlashGel™ System Sample Preparation Reagents
For use with the Lonza FlashGel System. FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.
Brand:  Lonza™ 57060
 View more versions of this product
Product Code. 11567865
  • 369.00 EUR / Pack of 2
Estimated Shipment: 07-02-2025
Description
  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA Recovery Buffer
Liquid
2 x 500 μL
Room Temperature