missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Latitude™ Midigel Chamber
Provides optimal running conditions for Latitude Precast Midigels
Brand: Lonza™ 56990
Includes: Acrylic buffer chamber and Acrylic safety cover with attached power supply leads.
Description
- Chamber has a red/orange background for easy sample loading
- Compact buffer compartment minimizes buffer usage
Note:this unit is designed for use with Latitude Precast Gels; casting accessories are not available
Specifications
Latitude™ Midigel Chamber | |
Acrylic | |
11.8 to 13.5 oz. | |
1 Year | |
9.4 in. | |
4.6 in. | |
2.7 in. | |
250 VDC | |
200 mA |
Acrylic buffer chamber and Acrylic safety cover with attached power supply leads. | |
55°C | |
350 to 400 mL | |
9.4 x 4.6 x 2.7 in. (24 x 11.8 x 7 cm) | |
24 cm | |
11.8 cm | |
7 cm | |
15 w |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction