missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ PAGEr™ Gold 4 to 20%, Tris-Glycine, Mini, Protein Gel, 16-well
Separation Range: 5 to 200 kd; Separation Type: Denaturing, Native; For Use With: CBS Scientific MGV System
Brand: Lonza™ 59517
This item is not returnable.
View return policy
Description
PAGEr Gold Precast Gels are easy-to-use protein minigels that offer sharp resolution and consistent protein transfer. Choose from multiple options for denatured or native protein separation over a wide molecular weight range, in an array of well configurations in both 9 × 10cm (see BMA58517 through BMA58524) and 10 × 10cm sizes to fit most popular chambers.
- Polyacrylamide minigels cast with 4% stacking gel for vertical protein electrophoresis
- Provide easy, accurate separation of biomolecules over wide molecular-weight range
- Produce crisp, razor-sharp resolution 5kDa - 250kDa
- Tris-Glycine buffer - Traditional Laemmli separation
- Marked sample lanes for easy loading and a simple twist-open design
- 8+1- and 17-well configurations are compatible with multichannel pipetters
- 2D-well configuration is designed for antibody testing and two-dimensional electrophoresis using a 7cm IPG strip
- Fit most vertical minigel boxes
- Packaged in a unique easy-to-open gold plastic cassette
- Available with a variety of polyacrylamide concentrations and well configurations
Certifications
ISO 9001:2000
Specifications
Plastic | |
Clear | |
4 to 20% | |
0.55 cm | |
Gradient Page | |
5 to 200 kDa | |
16-well | |
Polyacrylamide | |
2° to 8°C | |
2D Well | |
3.9 in. | |
10 cm | |
3.9 in. | |
10 cm |
ISO 9001:2000 | |
CBS Scientific MGV System, Daiichi/ISS 2 and 6 System, FisherBiotech Fisher EC 120-2, FisherBiotech Fisher FB-VE 10-1, FisherBiotech Fisher FB-VE 12-1, Hoefer Mighty Small SE 250, Hoefer Mighty Small SE 260, Hoefer Mighty Small SE280, Novex XCell I and II, XCell SureLock Mini-Cell, PAGEr Minigel Chamber, Owl Separation Systems Penguin P8DS-1, Sigma Aldrich Mini Techware, Zaxis System 200 | |
Mini | |
Tris-Glycine | |
Protein | |
Denaturing, Native | |
2D Electrophoresis, Denaturing and Native Protein Electrophoresis, Western Blotting | |
10/Pk. | |
14 μL | |
3.9 in. | |
10 cm | |
3.9 in. | |
10 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction