missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ PAGEr™ Gold 10%, Tris-Glycine, Mini, Protein Gel, 16-well
Separation Range: 25 to 200 kd; Separation Type: Denaturing, Native; For Use With: Bio-Rad Ready Gel Cell
Brand: Lonza™ 58514
This item is not returnable.
View return policy
Description
- Polyacrylamide minigels cast with 4% stacking gel for vertical protein electrophoresis
- Provide easy, accurate separation of biomolecules over wide molecular-weight range
- Produce crisp, razor-sharp resolution
- Tris-Glycine buffer - Traditional Laemmli separation
- Marked sample lanes for easy loading and a simple twist-open design
- Gel dimensions: 9 × 10cm
- 2D-well configuration is designed for antibody testing and two-dimensional electrophoresis using a 7cm IPG strip
- Fit most vertical minigel boxes
- Packaged in a unique easy-to-open gold plastic cassette
- Available with a variety of polyacrylamide concentrations and well configurations
Specifications
Plastic | |
10% | |
0.49 cm | |
Homogeneous Page | |
25 to 200 kDa | |
16-well | |
Polyacrylamide | |
2° to 8°C | |
2D Well | |
3.5 in. | |
9 cm | |
3.9 in. | |
10 cm |
Biometra Mini V, Bio-Rad Mini Protean Tanks, Bio-Rad Ready Gel Cell, CBS Scientific MGV System, FisherBiotech Fisher EC 120-2, Hoefer Mighty Small SE 250, Hoefer Mighty Small SE 260, Hoefer Mighty Small SE280, Novex XCell I and II, XCell SureLock Mini-Cell, PAGEr Minigel Chamber, Sigma Aldrich Mini Techware | |
Mini | |
Tris-Glycine | |
Protein | |
Denaturing, Native | |
2D Electrophoresis, Denaturing and Native Protein Electrophoresis, Western Blotting | |
10/Pk. | |
14 μL | |
3.5 in. | |
9 cm | |
3.9 in. | |
10 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction