Learn More
Lonza™ Reliant™ Precast RNA Gels
Formulated for separation of RNA of sizes from 0.25 to at least 10kb. Reliant™ Precast RNA Gels are ready-to-use RNase free agarose gels suitable for a variety of RNA applications including Northern blotting and analysis of RNA transcripts.
449.00 EUR
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
11537825
|
Lonza™
54922 |
Multiwell |
449.00 EUR
Pack of 20 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
11577835
|
Lonza™
54948 |
Multiwell |
449.00 EUR
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
Description
- Ready to use
- RNase free
- Free of denaturants, eliminating problems associated with staining gels containing formaldehyde
- Suitable for a variety of applications, including Northern blotting and analysis of RNA transcripts
- Compatible with most common mini chambers
- Available with a separate container of glyoxal buffer
Specifications
1.25% | |
Multiwell | |
Precast Gel |