Learn More
SeaKem™ HE Agarose, Lonza™
  • For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
  • High EEO (0.23 –0.26)
  • Standard gelling temperature agarose
235.00 EUR - 785.00 EUR
Specifications
Chemical Name or Material SeaKem HE Agarose
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11537775
View Documents
Lonza
50021
25 g
235.00 EUR
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
11527775
View Documents
Lonza
50020
125 g
785.00 EUR
Each
Estimated Shipment: 04-02-2025
to see stock.
Specifications
SeaKem HE Agarose