Learn More
SeaKem™ HGT Agarose, Lonza™
- Used in counterimmunoelectrophoresis and crossed immunoelectrophoresis
- High gelling temperature (42°C ± 1.5°C)
- High gel strength (≥1000g/cm2) and clarity
Brand: Lonza 50041
This item is not returnable.
View return policy
Description
Specifications
SeaKem HGT Agarose | |
25 g |