Learn More
SeaPlaque™ Agarose, Lonza™
  • For preparative DNA and RNA electrophoresis
  • Molecular biology grade
  • ≤0.10 EEO
  • Low melting temperature argarose (26°–30°C Gelling temperature)
  • DNase/RNase-free
559.00 EUR - 1820.00 EUR
Specifications
Chemical Name or Material SeaPlaque™ Agarose
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11587785
View Documents
Lonza
50101
25 g
559.00 EUR
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
11577785
View Documents
Lonza
50100
125 g
1820.00 EUR
125g
Estimated Shipment: 19-02-2025
to see stock.
Specifications
SeaPlaque™ Agarose