Learn More
SPEX™ Low Binding Silica Grinding Beads
Product Code. 15380422 Shop All SPEX Certiprep Products
Click to view available options
Product Type:
400 μm beads
800 μm beads
Unit Size:
200g
Product Code. 15380422
Brand: SPEX™ 2162
This item requires a price update prior to being ordered.
Request a Price for this item here.
SPEX™ Low Binding Silica Grinding Beads are acid washed silica grinding beads that are chemically treated to keep samples from binding to titer plate.
  • Used for lysing dilute samples of cells in the Geno/Grinder
  • Treated to remove fine particles and contaminates
  • Coated to reduce non-specific binding of nucleic acids and proteins
  • Bead size approximately 400 μm and 800 μm
  • Compatible with all SPEX™ SamplePrep Titer Plates and Vial Sets
Specifications
Product Type 800 μm beads
Material Silica
Packaging Quantity 200 g bottle