Learn More
MP Biomedicals Low Iodine Diet, Fortified With Vitamins, Pellet
Vitamin fortified diet, low in iodine.
Brand:  MP Biomedicals 0290220420
 View more versions of this product
Product Code. 11445592
  • 1619.68 EUR / 20kg
Explore more special offers
Description
Description
  • This diet is low in iodine but fortified with other vitamins
  • Ideal for research on iodine deficiency diets
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
20 kg
Room Temperature
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.