Learn More
Thermo Scientific™ Luer-Tip, Gas-Tight Manual Syringes for GC Systems
Product Code. 10371195 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
5 μL
10 μL
1000 μL
10 mL
Needle Gauge:
23s G
26 G
26s G
Length (Metric):
57 mm
80 mm
85 mm
Unit Size:
Each
For Use With (Equipment):
Gas Chromatography Systems
TriPlus RSH Autosampler
TriPlus RSH, TriPlus 100LS Autosamplers
Product Code. 10371195
Brand: Thermo Scientific™ 36502025
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get a gas-tight syringe for manual injections. This syringe features a Luer-Tip connection and is available in a variety of sizes.
Get a gas-tight syringe for manual injections in your GC system. This syringe features a Luer-Tip connection and is available in a variety of sizes. Made of quality materials, they are designed for easy, accurate, precise and reproducible injections.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 26 G
Length (English) 3.15 in.
Length (Metric) 80 mm
Volume (Metric) 5 μL
Needle Point Style Cone
Connection Type Fixed