missing translation for 'onlineSavingsMsg'
Learn More

LY 341495 disodium salt, Tocris Bioscience™

Product Code. 18085342
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective group II mGlu antagonist; disodium salt of LY 341495 (Cat. No. 1209)

Highly potent and selective group II metabotropic glutamate receptor antagonist. Disodium salt of LY 341495 (Cat. No. 1209).

Chemical Identifiers

Molecular Formula C20H17NNa2O5
Synonym (2S)-2-Amino-2-[(1S, 2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid disodium salt

Specifications

Quantity 1 mg
Formula Weight 397.33
Percent Purity >97%
Chemical Name or Material LY 341495 disodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'