missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 341495

Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
Highly potent and selective group II metabotropic glutamate receptor antagonist (Ki/IC50 values are 2.3, 1.3, 173, 990, 6800, 8200 and 22000 nM for human mGlu2, mGlu3, mGlu8, mGlu7a, mGlu1a, mGlu5a and mGlu4a receptors respectively). Readily brain penetrant and active in vivo.
Disodium salt (Cat. No. 4062) also available.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 353.37 |
| Purity | >98% |
| Molecular Weight (g/mol) | 353.374 |
| Chemical Name or Material | LY 341495 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction