missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab MacConkey Agar ( Medium H Eur. Pharm./HP ) ( Mac Conkey Agar Nº3 )
Click to view available options
:
MacConkey Agar; Color: Red
Description
- AOAC/BAM/COMPF/EP/HP/ISO/SMWW/USP
- Selective and differential medium
- Used in the detection, isolation and enumeration of Salmonella and coliforms in clinical specimens
- According to the Pharmacopoea Harmonized Methodology and in foodstuffs specimens according to ISO 21150 standard.
Specifications
Specifications
| Certifications/Compliance | ISO 21150 |
| Color | Red |
| Form | Solid |
| Media Type | MacConkey Agar |
| Product Type | Selective and Differential Medium |
| Target Organism Class | Coliforms, Gram-negative Bacteria |
| Formulation | Pancreatic Digest of Gelatin 17.00 g/L, Meat Peptone 1.50 g/L, Casein Peptone 1.50 g/L, Lactose Monohydrate 10.00 g/L, Bile Salts 1.50 g/L, Sodium Chloride 5.00 g/L, Neutral Red 0.030 g/L, Crystal Violet 0.001 g/L, Agar 15.00 g/L |
| For Use With (Application) | Isolation and Enumeration of Salmonella and Coliforms in Clinical Specimens |
| Incubation Conditions | 30°C to 35°C |
| Incubation Time | 18 to 72 hr. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction