missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents

Product Code. 30054576 Shop All Applied Biosystems Products
Click to view available options
Product Type:
DNA Lysis/Binding Solution
Proteinase Digestion Buffer
RNA Binding Solution
RNA Wash Solution I Concentrate
Sequential Binding Beads
Sequential DNA Elution Buffer
Sequential DNA Wash Solution I
Sequential DNA/RNA Kit
Sequential DNase Buffer
Sequential DNase I
Sequential Proteinase K
Sequential RNA Elution Buffer
Unit Size:
0.4mL
120mL
16mL
24mL
300mL
32mL
40mL
462mL
4mL
6mL
80mL
Each
This item is not returnable. View return policy

Product Code. 30054576

Brand: Applied Biosystems™ A66593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples, when paired with a KingFisher purification system.

The MagMAX Sequential DNA/RNA kit and reagents, when paired with a KingFisher purification system, are an automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples. This innovative kit and standalone reagents eliminate the need to purchase and run separate DNA and RNA purification kits, streamlining your workflow and maximizing efficiency.

The MagMAX Sequential DNA/RNA kit and reagents, when paired with a KingFisher purification system, are an automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples. This innovative kit and standalone reagents eliminate the need to purchase and run separate DNA and RNA purification kits, helping streamline your workflow and maximizing efficiency.

Features of the MagMAX Sequential DNA/RNA Kit include:

  • Test utilization management—obtain more information using minimal sample volume to help preserve precious samples
  • Versatile workflow solutions—flexible workflow enables low- to high-throughput sample processing and various sample input volume formats
  • Easy identification—color-coded lid labels enable quick reagent identification, helping to minimize the risk of costly errors
  • Sequential DNA and RNA isolation—obtain high-quality genomic DNA and total RNA from the same blood or bone marrow sample
  • Compatible with KingFisher instruments—for a streamlined, automated workflow solution
  • Expand hematologic research—separate, ready-to-use DNA and RNA eluates for use in broad range of downstream applications
  • Available as stand-alone products—reagents available in larger volume bottles sold separately

Made for KingFisher purification systems

The MagMAX Sequential DNA/RNA Kit is compatible with the KingFisher Duo Prime, KingFisher Flex, and KingFisher Apex purification systems, allowing for a semi-automated workflow with a total turnaround time in about 2.5 hours. By isolating genomic DNA and total RNA from a single sample in one sequential workflow, the MagMAX Sequential DNA/RNA Kit simplifies nucleic acid purification for a broad range of molecular downstream applications, such as PCR and sequencing. This all-in-one kit is an excellent choice for researchers seeking an automated, efficient workflow that delivers reliable results from limited sample material.

Working with higher throughput sample numbers or input volumes?

The MagMAX Sequential DNA/RNA Kit reagents are also available as stand-alone products sold separately. These reagents come in larger volumes to facilitate high-throughput sample processing and accommodate higher sample input volumes. When purchased as a set of eleven stand-alone reagents, users will have sufficient MagMAX Sequential DNA/RNA reagent volumes to process 440 samples ranging from 200–500 μL each.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Format Reagent
Isolation Technology Magnetic Bead
Sample Type Whole Blood, Bone Marrow
For Use With (Equipment) KingFisher™ Duo Prime, Flex, or Apex purification systems
For Use With (Application) Molecular applications, including qPCR and sequencing
Nucleic Acid Type Genomic DNA, Total RNA, miRNA
Product Line MagMAX
Product Type RNA Wash Solution I Concentrate
Quantity 462 mL
Shipping Condition Room Temperature
Throughput Flexible
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.