Learn More
Kartell™ Magnetic Triangular Stirring Bars
Made from Polytetrafluoroethylene. Kartell™ 120° Magnetic Triangular Stirring Bars are ideal for dissolving solids and mixing sediments thanks to the scraping action. The angled surfaces create high turbulence even at low speeds.
Brand:  Kartell™ 0078100
Additional Details : Weight : 0.10000kg
Product Code. 10346061
  • 45.25 EUR / Pack of 10
Estimated Shipment: 22-07-2024
Description
  • Cuneiform
  • Angled surface
  • Suitable for foodstuff
  • Magnet PTFE coated
Specifications
Specifications
7.39x25.x32mm
PTFE (Polytetrafluoroethylene)