Learn More
Invitrogen™ MagniSort™ Human CD45 Depletion Kit
Designed to deplete CD45+ cells from human lysed whole blood or peripheral blood mononuclear cells, utilizing a biotinylated Anti-Human CD45 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6802-74
536.02 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
Contains Sodium azide
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes:
- MagniSort Human CD45 Biotin A : 200 tests, 20μL/test
- MagniSort Negative Selection Beads B
Description
- CD45+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting
- After depletion, the purity of selected cells can be verified by staining with Anti-Human CD45, clone 2D1
Specifications
CD45+ | |
200 tests | |
Human |
Cell Depletion Kit | |
200 tests |