Learn More
Value Plastics Male Luer Slip to Plug Adapters
Seal luer ports with a secure and leak-proof connection
Brand: Value Plastics 50120-82
This item is not returnable.
View return policy
Description
Luer fittings allow for a simple yet secure connection. Ideal for capping off fluid transfer system. Quickly connect and disconnect with slip luer designSpecifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP | |
Male Luer Slip, Plug, Closed at Grip | |
Polypropylene |