Learn More
Emma Safety Footwear Mammoet M1807 XD Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Heat and cold insulated
- Fire-retardant laces
- Waterproof Oil Nubuck construction
- Hydro-Tec™ Sanitized Silver
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Mammoet M1807 XD |
| Liner Material | Hydro-Tec™ Sanitized Silver / Sympatex™ |
| Material | Oiled Nubuck |
| Size | 49 |
| Toe Material | Steel |