missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14049-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in the maturation of Asn-linked oligosaccharides. Progressively trim alpha-1, 2-linked mannose residues from Man(9)GlcNAc(2) to produce Man(5)GlcNAc(2).Specifications
MAN1A2 | |
Polyclonal | |
Unconjugated | |
MAN1A2 | |
Alpha 1,2 mannosidase IB, MAN1A2, MAN1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10905 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60476 | |
MAN1A2 | |
MAN1A2 Fusion Protein Ag5240 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |