missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, ExpertQ, Scharlab

Product Code. 11348424
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 10034-96-5
Quantity 1 kg
Molecular Formula H2MnO5S
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES O.[Mn++].[O-]S([O-])(=O)=O
IUPAC Name manganese(2+) hydrate sulfate
Molecular Weight (g/mol) 169.01
Grade ExpertQ
compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate monohydrate, reagent grade, ACS, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.