Learn More
Watson-Marlow Manifold Tubing - Silicone
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand: Watson-Marlow 983.0185.000
This item is not returnable.
View return policy
Description
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
7/100 in. | |
Silicone | |
0.8 mm | |
1.85mm | |
Green/Green |
1.85 mm | |
0.03 in. | |
400DM2 and 400DM3 pump heads and multichannel cartridge | |
0.40 to 28 (four roller pump), 0.29 to 20 (eight roller pump)mL/min. | |
Pump Tubing |