Learn More
Watson-Marlow Manifold Tubing - Silicone
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 983.0185.000
 View more versions of this product
Product Code. 13430897
  • 342.00 EUR / Pack of 6
Estimated Shipment: 12-03-2025
Description
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7/100 in.
Silicone
0.8 mm
1.85mm
Green/Green
1.85 mm
0.03 in.
400DM2 and 400DM3 pump heads and multichannel cartridge
0.40 to 28 (four roller pump), 0.29 to 20 (eight roller pump)mL/min.
Pump Tubing