Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
- Simply grip pliers around top of seal, squeeze and pry upward to remove
Specifications
Specifications
| Description | Decapping Pliers |
| No. per Pack | 1 Ea. |
| Type | Manual Decapping Plier |
| For Use With (Equipment) | National Vial, Chromacol Vial, Pry Seal from Crimp Top, Vialsry Seals from Crimp Top Vial |
| Size | 30 mm |
| For Use With (Application) | Industrial Chromatography |
| Product Line | National |