Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
11 mm Crimp Vials
13 mm Crimp Vials
20 mm Crimp Vials
8 mm Crimp Vials
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
Description
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifications
Specifications
| Type | Manual Decrimper |
| For Use With (Equipment) | 20 mm Crimp Vials |
| Material | Stainless Steel/Aluminum |
| Product Line | SureSTART Level 2 |
Product Content Correction
Product Title
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials >
Spot an opportunity for improvement?Share a Content Correction