Learn More
SI Analytics™ Cannon-Fenske Reverse Flow Viscometer
Product Code. 10538981 Shop All SI Analytics Products
Click to view available options
Viscosity Range:
20 to 100 cSt
3 to 15 cSt
7 to 35 cSt
Approximate Constant:
0.0215K
0.035K
0.1K
Size:
0.63mm Diameter
0.78mm Diameter
1.02mm Diameter
Unit Size:
Each
Product Code. 10538981
Brand: SI Analytics™ 285403042
Only null left
Add to basket
Add to basket
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Cannon-Fenske Reverse Flow Viscometer includes three ring marks since they can only be checked by means of manual measurements.
  • Suitable for transparent liquids manual measurement, opaque liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • Calibrated
  • With two constants
  • With three ring marks
  • Only for manual measurements
  • Filling quantity of 12mL
  • Overall length of 295mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 7 to 35 cSt
Calibrated Yes
Description Capillary No. 150
Approximate Constant 0.035K
ASTM Numbers D 446
Certifications/Compliance ISO/DIS 3105, ASTMD 2515, ASTM D 446, NF T 60 - 100
Includes Ring Marks and 2 Constants
For Use With (Application) Only for manual measurement
Size 0.78mm Diameter
Volume (Metric) 12 mL