Learn More
Notrax Marble Sof-Tyle™ Marbleized Rubber Anti-Fatigue Mat
Product Code. p-7227319
Click to view available options
Dimensions (L x W):
91 x 60 cm
150 x 91 cm
2286 x 60 cm
2286 x 91 cm
2286 x 122 cm
Unit Size:
Each
Product Code. 17732158
Brand: Notrax 470S2436GY
Only null left
Add to basket
Add to basket
Heavy duty anti-fatigue mat with a smooth rubber surface with marbleized pattern laminated with UniFusion™ technology and RedStop™ slip-resistant backing. Combines style with exceptional comfort and durability. Ideal for commercial, laboratory, and tertiary environments.
  • Smooth rubber top surface embedded with a marbleized pattern
  • Rubber naturally provides traction to resist slips and falls
  • Long wearing top surface laminated on to a resilient microcellular vinyl base for maximum durability coupled with fatigue relief
  • Resistant to most industrial fluids and chemicals
  • Featured with RedStop™ slip resistant backing to prevent mats from shifting
  • Bevelled edges on all four sides to minimize risk of tripping
  • UniFusion™ technology, engineered to resist in the harshest work environments
Specifications
Dimensions (L x W) 91 x 60 cm
Color Gray
Length (English) 36 in.
Length (Metric) 91 cm
Material PVC on Microcellular Vinyl Base
Type Industrial and Commercial Matting
Width (Metric) 60 cm