Learn More
Watson-Marlow Marprene Double Segment Manifold Tubing
Marprene Double Segment Manifold Tubing is made from a long life thermoplastic elastomer with resistance to a wide range of chemicals.
Brand: Watson-Marlow 979.0129.000
This item is not returnable.
View return policy
Description
- Marprene is a long life thermoplastic elastomer with resistance to a wide range of chemicals.
- It lasts at least 10 times as long as tubing in other materials.
- Long-life tubing means stoppages for a tube change are rare events, easily fitted in with general maintenance.
- Marprene has wide chemical compatibility, and is highly resistant to oxidizing agents such as ozone, peroxides and sodium hypochlorite.
- Marprene is beige and opaque to visible and ultraviolet light, with low permeability to oxygen, carbon dioxide and nitrogen.
- It meets USDA standards for food handling.
Specifications
Double Segment Tubing | |
1.29 mm | |
1.5 ft. | |
0.03 in. | |
Grey/Grey |
0.05 in. | |
Marprene | |
0.46 m | |
0.8 mm |