Learn More
Masterflex™ I/P™ End Bell Assembly
Click to view available options
Material:
Polycarbonate, Cold-rolled Steel
Polycarbonate, Stainless Steel
Unit Size:
Each
Description
- Assemblies are made from high-performance Polyphenylene sulfide and stainless steel
Specifications
Specifications
| For Use With | For Masterflex I/P 73 standard pump heads |
| Material | Polycarbonate, Stainless Steel |
| Quantity | 1 Each |
| Type | End Bell Assembly |