missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MASTL, GST Tag Recombinant Protein
This MASTL Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A30978
This item is not returnable.
View return policy
Description
- This MASTL recombinant human protein (full length) was expressed in insect cells
- MASTL, or microtubule associated serine/threonine kinase-like, belongs to the MAST family and AGC Ser/Thr protein kinase group
- MASTL promotes mitotic progression and cell cycle reentry after DNA damage
- Loss of MASTL leads to defects in chromosome condensation, separation, and other aspects of mitotic progression, which is primarily mediated by inhibiting PP2A/B55 delta
- Overexpression of MASTL is correlated with cancer progression, poor patient survival, and tumor recurrence in various cancers
- MASTL recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_001165774 | |
123.6 kDa | |
MAST Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
10 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Baculovirus | |
Recombinant, Full Length |